missing translation for 'onlineSavingsMsg'
Learn More

Donepezil Benzyl Bromide (Donepezil Impurity), TRC

Product Code. 30385361
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385361 5 mg
30380233 50 mg
30371021 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385361 Supplier Toronto Research Chemicals Supplier No. TRCD5317405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844694-85-5
Molecular Formula C31H36BrNO3
Molecular Weight (g/mol) 550.53
Synonym Piperidinium, 4-[(2,3-dihydro-5,6-dimethoxy-1-oxo-1H-inden-2-yl)methyl]-1,1-bis(phenylmethyl)-, bromide (1:1), Piperidinium, 4-[(2,3-dihydro-5,6-dimethoxy-1-oxo-1H-inden-2-yl)methyl]-1,1-bis(phenylmethyl)-, bromide (9CI), 4-[(2,3-Dihydro-5,6-dimethoxy-1-oxo-1H-inden-2-yl)methyl]-1,1-bis(phenylmethyl)piperidinium bromide (1:1), 1,1-Dibenzyl-4-[(5,6-dimethoxy-1-oxo-2,3-dihydro-1H-inden-2-yl)methyl]piperidinium bromide, Donepezil quaternary salt, Donepezilbenzyl bromide
SMILES [Br-].COc1cc2CC(CC3CC[N+](Cc4ccccc4)(Cc5ccccc5)CC3)C(=O)c2cc1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C31H36NO3.BrH/c1-34-29-19-26-18-27(31(33)28(26)20-30(29)35-2)17-23-13-15-32(16-14-23,21-24-9-5-3-6-10-24)22-25-11-7-4-8-12-25;/h3-12,19-20,23,27H,13-18,21-22H2,1-2H3;1H/q+1;/p-1
Percent Purity >95%
Chemical Name or Material Donepezil Benzyl Bromide (Donepezil Impurity)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.