missing translation for 'onlineSavingsMsg'
Learn More

Donepezil-13C3 Hydrochloride, TRC

Product Code. 30382145
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382145 5 mg -
30380530 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382145 Supplier Toronto Research Chemicals Supplier No. TRCD5317555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246814-90-3
Molecular Formula 13C3 C21 H29 N O3 . Cl H
Molecular Weight (g/mol) 418.93
Synonym 2,3-Dihydro-5,6-dimethoxy-2-[(1-phenylmethyl-4-piperidinyl)methyl]-1H-inden-1-one-1,2,3-13C3 hydrochloride, 2,3-Dihydro-5,6-dimethoxy-2-[(1-benzyl-4-piperidinyl)methyl]-1H-inden-1-one-1,2,3-13C3 hydrochloride, Donepezil-13C3 hydrochloride, Donepezil-13C3 hydrochloride (1H-inden-1-one-1,2,3-13C3)
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride
SMILES Cl.COc1cc2[13CH2][13CH](CC3CCN(Cc4ccccc4)CC3)[13C](=O)c2cc1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H29NO3.ClH/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18;/h3-7,14-15,17,20H,8-13,16H2,1-2H3;1H/i13+1,20+1,24+1;
Formula Weight 418.2015
Chemical Name or Material Donepezil-13C3 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.