missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir sodium salt, Thermo Scientific™

Product Code. p-7227409 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17700709

Brand: Thermo Scientific Acros 469770010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Dolutegravir sodium salt
CAS 1051375-19-9
Quantity 1 g
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C20 H18 F2 N3 Na O5
Physical Form Powder or Crystals
Color White to Light-yellow
Purity ≥98.5%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.