missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir sodium salt, Thermo Scientific™

Product Code. 17710709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710709 250 mg 250mg
17700709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710709 Supplier Thermo Scientific Acros Supplier No. 469772500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Dolutegravir sodium salt
CAS 1051375-19-9
Quantity 250 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C20 H18 F2 N3 Na O5
Physical Form Powder or Crystals
Color White to Light-yellow
Purity ≥98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.