missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir, MedChemExpress

Product Code. 16451329
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451329

Brand: Medchem Express HY13238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dolutegravir (S/GSK1349572) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer. Dolutegravir (S/GSK1349572) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM).

Dolutegravir (S/GSK1349572) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer. Dolutegravir (S/GSK1349572) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM).

Specifications

Chemical Name or Material Dolutegravir
Physical Form Solid
CAS 1051375-16-6
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C20H19F2N3O5
For Use With (Application) COVID-19-anti-virus
Synonym S/GSK1349572
Solubility Information DMSO : 10 mg/mL (23.84 mM; Need ultrasonic and warming)
SMILES FC1=CC(F)=C(C=C1)CNC(C2=CN(C3=C(C2=O)O)C[C@@]([H])(N4C3=O)OCC[C@H]4C)=O
Molecular Weight (g/mol) 419.38
Formula Weight 419.38
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.