missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir intermediate-1, TRC

Product Code. 30380209
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380209 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380209 Supplier Toronto Research Chemicals Supplier No. TRCD528812250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid
CAS 1335210-23-5
Molecular Formula C13 H17 N O8
Quantity 250 mg
Synonym 2,5-Pyridinedicarboxylic acid, 1-(2,2-dimethoxyethyl)-1,4-dihydro-3-methoxy-4-oxo-, 2-methyl ester, 1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 1-[2,2-Bis(methyloxy)ethyl]-5-(methyloxy)-6-[(methyloxy)carbonyl]-4-oxo-1,4-dihydro-3-pyridinecarboxylic acid, 1-(2,2-Dimethoxyethyl)-1,4-dihydro-3-methoxy-4-oxo-2,5-pyridinedicarboxylic Acid 2-Methyl Ester, 2,5-Pyridinedicarboxylic acid, 1-(2,2-dimethoxyethyl)-1,4-dihydro-3-methoxy-4-oxo-, 2-methyl ester, 1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 1-[2,2-Bis(methyloxy)ethyl]-5-(methyloxy)-6-[(methyloxy)carbonyl]-4-oxo-1,4-dihydro-3-pyridinecarboxylic acid, DHPA, GSK 2598836A, Dolutegravir related
InChI Formula InChI=1S/C13H17NO8/c1-19-8(20-2)6-14-5-7(12(16)17)10(15)11(21-3)9(14)13(18)22-4/h5,8H,6H2,1-4H3,(H,16,17)
SMILES COC(CN1C=C(C(=O)O)C(=O)C(=C1C(=O)OC)OC)OC
IUPAC Name 1-(2,2-dimethoxyethyl)-5-methoxy-6-methoxycarbonyl-4-oxopyridine-3-carboxylic acid
Molecular Weight (g/mol) 315.28
Formula Weight 315.0954
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.