missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOLPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553944
This item is not returnable.
View return policy
Description
Immunogen sequence: TQEVLTPLFP RIAAWPVSEF FLIRDTSLIP NVLWFEYTVT RAEARNRQRK LGTKL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Required for efficient N-glycosylation. Necessary for maintaining optimal levels of dolichol-linked oligosaccharides. Hydrolyzes dolichyl pyrophosphate at a very high rate and dolichyl monophosphate at a much lower rate. Does not act on phosphatidate.
Specifications
| DOLPP1 | |
| Polyclonal | |
| Unconjugated | |
| Dolpp1 | |
| 0610011H20Rik; AB030189; dolichyl pyrophosphate phosphatase 1; Dolichyldiphosphatase 1; DOLPP1; linked to Surfeit genes in Fugu rubripes 2; LSFR2; Protein 2-23; Unknown (protein for IMAGE:8120927); zgc:101585 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57171 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86YN1 | |
| Dolpp1 | |
| Recombinant protein corresponding to Human DOLPP1. Recombinant protein control fragment (Product #RP-91024). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction