missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Dolly, steel, 150kg capacity

Brand:  Key Industrial Equipment™ 1252O054

Encompass

Product Code. 12817828

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Steel dolly with all round PVC buffer strip to protect walls and doorways. Non-slip PVC deck surface.
  • Dimensions (H x W x L): 135mm x 480mm x 830mm
  • Fitted with two fixed and two swivel castors
  • Load capacity: 150kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.