missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOLK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584786
This item is not returnable.
View return policy
Description
Immunogen sequence: YQNAKRSSSE SKKHQAPTIA RKY Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
Involved in the synthesis of the sugar donor Dol-P-Man which is required in the synthesis of N-linked and O-linked oligosaccharides and for that of GPI anchors.
Specifications
| DOLK | |
| Polyclonal | |
| Unconjugated | |
| Dolk | |
| CDG1M; DK; DK1; dolichol kinase; dolichol kinase 1; DOLK; KIAA1094; SEC59; SEC59 homolog; TMEM15; Transmembrane protein 15; UNQ2422/PRO4980 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPQ8 | |
| Dolk | |
| Recombinant protein corresponding to Human DOLK. Recombinant protein control fragment (Product #RP-108543). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction