missing translation for 'onlineSavingsMsg'
Learn More

Dolastatin 10, MedChemExpress

Product Code. p-200101591
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491529

Brand: Medchem Express HY15580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dolastatin 10 (DLS 10) is a potent antimitotic peptide that inhibits tubulin polymerization.

Dolastatin 10 (DLS 10) is a potent antimitotic peptide that inhibits tubulin polymerization.

Specifications

Chemical Name or Material Dolastatin 10
Physical Form Solid
CAS 110417-88-4
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C42H68N6O6S
For Use With (Application) Cancer-programmed cell death
Synonym DLS 10 NSC 376128
Solubility Information DMSO : ≥ 100 mg/mL (127.37 mM)
SMILES O=C([C@H](C)[C@H]([C@]1([H])CCCN1C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@H](NC([C@@H](N(C)C)C(C)C)=O)C(C)C)=O)C)=O)OC)N[C@@H](CC2=CC=CC=C2)C3=NC=CS3
Molecular Weight (g/mol) 785.09
Formula Weight 785.09
Percent Purity 97.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.