missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dolastatin 10, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Dolastatin 10 (DLS 10) is a potent antimitotic peptide that inhibits tubulin polymerization.
Specifications
Specifications
| Chemical Name or Material | Dolastatin 10 |
| Physical Form | Solid |
| CAS | 110417-88-4 |
| Color | Off-White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C42H68N6O6S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | DLS 10 NSC 376128 |
| Solubility Information | DMSO : ≥ 100 mg/mL (127.37 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction