missing translation for 'onlineSavingsMsg'
Learn More

Dolasetron, TRC

Product Code. 30370818
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370818 10 mg
30387938 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370818 Supplier Toronto Research Chemicals Supplier No. TRCD52850010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115956-12-2
Molecular Formula C19 H20 N2 O3
Molecular Weight (g/mol) 324.37
Synonym 1H-Indole-3-carboxylic acid, octahydro-3-oxo-2,6-methano-2H-quinolizin-8-yl ester, stereoisomer, 1H-Indole-3-carboxylic acid, octahydro-3-oxo-2,6-methano-2H-quinolizin-8-yl ester, (2α,6α,8α,9aβ)-, 2,6-Methano-2H-quinolizine, 1H-indole-3-carboxylic acid deriv., Anemet, Dolasetron, MDL 73147
SMILES O=C(O[C@@H]1C[C@@H]2C[C@H]3C[C@H](C1)N2CC3=O)c4c[nH]c5ccccc45

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H20N2O3/c22-18-10-21-12-5-11(18)6-13(21)8-14(7-12)24-19(23)16-9-20-17-4-2-1-3-15(16)17/h1-4,9,11-14,20H,5-8,10H2/t11-,12+,13-,14-
Formula Weight 324.1474
Chemical Name or Material Dolasetron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.