missing translation for 'onlineSavingsMsg'
Learn More

Dolasetron Mesylate hydrate, MedChemExpress

Product Code. 16491436
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491436 200 mg 200mg
16428446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491436 Supplier Medchem Express Supplier No. HYB0750B
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dolasetron Mesylate hydrate (MDL-73147EF hydrate) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.

Dolasetron Mesylate hydrate (MDL-73147EF hydrate) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.

Specifications

Chemical Name or Material Dolasetron Mesylate hydrate
CAS 878143-33-0
Color White
Purity Grade Notes Research
Molecular Formula C20H26N2O7S
Quantity 200 mg
Synonym MDL-73147EF hydrate
Solubility Information DMSO : 250 mg/mL (570.14 mM; Need ultrasonic)
SMILES O=C(C1=CNC2=C1C=CC=C2)O[C@@H]3C[C@@](CC4C5)([H])[N@](CC4=O)[C@@]5([H])C3.O=S(C)(O)=O.O
Molecular Weight (g/mol) 438.49
Formula Weight 438.49
Percent Purity 98.73%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.