missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dolasetron, MedChemExpress
Description
Dolasetron(MDL-73147) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.
Specifications
Specifications
| Chemical Name or Material | Dolasetron |
| CAS | 115956-12-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H20N2O3 |
| Quantity | 5 mg |
| Synonym | MDL-73147 |
| Solubility Information | DMSO : ≥ 300 mg/mL (924.87 mM) |
| SMILES | O=C(C1=CNC2=C1C=CC=C2)O[C@@H]3C[C@@](CC4C5)([H])[N@](CC4=O)[C@@]5([H])C3 |
| Molecular Weight (g/mol) | 324.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?