Learn More
Invitrogen™ DOK5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103194
Product Code. 16669644
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total DOK5.
The protein encoded by this gene is a member of the DOK family of membrane proteins, which are adapter proteins involved in signal transduction. The encoded protein interacts with phosphorylated receptor tyrosine kinases to mediate neurite outgrowth and activation of the MAP kinase pathway. In contrast to other DOK family proteins, this protein does not interact with RASGAP.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DOK5
Polyclonal
Unconjugated
DOK5
2700055C10Rik; C20orf180; Docking protein 5; Dok5; Downstream of tyrosine kinase 5; insulin receptor substrate 6; IRS6; IRS-6; MGC16926; Protein dok-5; RGD1562846
Rabbit
Affinity chromatography
RUO
55816, 76829
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q91ZM9, Q9P104
DOK5
A synthesized peptide derived from human DOK5(Accession Q9P104), corresponding to amino acid residues T273-H306.
100 μL
Primary
Human, Mouse