missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549841
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DOK3 protein.
DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function.
Specifications
| DOK3 | |
| Polyclonal | |
| Unconjugated | |
| DOK3 | |
| AI450713; Docking protein 3; Dok3; Dokl; DOK-L; Dok-like protein; downstream of tyrosine kinase 3; FLJ22570; FLJ39939; p62(dok)-like protein; p62Dok-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27261, 79930 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7L591, Q9QZK7 | |
| DOK3 | |
| Synthesized peptide derived from internal of human DOK3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction