Learn More
Invitrogen™ DOK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589616
Description
Immunogen sequence: MGDGAVKQGF LYLQQQQTFG KKWRRFGASL YGGSDCALAR LELQEGPEKP RRCEAARKVI RLSDCLRVAE AGGEASSPRD TSAFFLETKE RLYLLAAPAA ERGDWVQAIC LLAFPGQRKE LSGPEGKQSR PCMEENELYS SAVTVGPHKE FAVTMRPTEA SERCHLRGSY TLRAGESALE LWGGPEPGTQ LYDWPYRFLR RFGRDKVTFS FEAGRRCVSG EGNFEFETRQ GNEIFLALEE AISAQKNAAP ATPQPQPATI; Positive Samples: THP-1, Mouse lung.
The protein encoded by this gene is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. This encoded protein binds p120 from CML cells.
Specifications
| DOK2 | |
| Polyclonal | |
| Unconjugated | |
| DOK2 | |
| docking protein 2; docking protein 2, 56kD; docking protein 2, 56kDa; Dok2; Dok-2; DokR; Dok-R; dok-related protein; Downstream of tyrosine kinase 2; Frip; IL-Four Receptor Interacting Protein; IL-four receptor-interacting protein; LOW QUALITY PROTEIN: docking protein 2; docking protein 2; p56 dok-2; p56(dok-2); p56DOK; p56dok-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13449, 9046 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60496, O70469 | |
| DOK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DOK2 (NP_0039652). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.