missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527464
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (88%), Rat (89%), Pig (93%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DOK1 (P62dok, Docking protein 1) is believed to be a mainly cytoplasmic adaptor protein which down-regulates mitogen-activated protein kinase activation, inhibits cell proliferation and transformation, and promotes cell spreading and cell migration. DOK1 is a major substrate for many tyrosine kinases including c-kit, v-abl, vFPS, EGF, and PDGF. Upon phosphorylation by kinases, DOK1 forms a complex with ras GTPase-activating protein. Phosphorylation on tyrosine residues by the insulin receptor kinase results in the negative regulation of the insulin signaling pathway. DOK1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs.
Specifications
| DOK1 | |
| Polyclonal | |
| Unconjugated | |
| DOK1 | |
| AW557123; Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Dok; DOK 1; DOK1; Dok-1; Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; MGC117395; MGC138860; p240; p62 dok; p62(dok); p62DOK; pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1796 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99704 | |
| DOK1 | |
| Recombinant protein encompassing a sequence within the center region of human p62Dok. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction