missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00
Specifications
| Antigen | DOK1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
DOK1 Polyclonal specifically detects DOK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| DOK1 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| docking protein 1, docking protein 1 (downstream of tyrosine kinase 1), docking protein 1, 62kD (downstream of tyrosine kinase 1), docking protein 1, 62kDa (downstream of tyrosine kinase 1), Downstream of tyrosine kinase 1, MGC117395, MGC138860, p62(dok), P62DOK, pp62 | |
| DOK1 | |
| IgG | |
| Affinity Purified | |
| 62 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| AAC51127 | |
| 1796 | |
| Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human DOK1. The immunogen is located within amino acids 410 - 460 of DOK1. Amino Acid Squence: PQGPAFPEPGTATGS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title