missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG1/TMEM16A Rabbit anti-Human, Clone: DG1/2564R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07652-100ug
This item is not returnable.
View return policy
Description
DOG1/TMEM16A Monoclonal specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| DOG1/TMEM16A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
| A recombinant human DOG1/TMEM16A protein fragment (around aa 2-101) (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DG1/2564R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 55107 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction