missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG1/TMEM16A Antibody (SPM580) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44379
This item is not returnable.
View return policy
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DOG1/TMEM16A | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55107 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SPM580 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.25 - 0.5 μg/mL | |
| ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
| Recombinant human canine1/TMEM16A protein (Uniprot: Q5XX6) | |
| 7 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction