missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG1/TMEM16A Antibody (Novartis patent anti-TMEM16A) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-27985JF635
This item is not returnable.
View return policy
Description
DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DOG1/TMEM16A | |
| Monoclonal | |
| Janelia Fluor 635 | |
| ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A | |
| ANO1/ TMEM16A | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Novartis patent anti-TMEM16A | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 55107 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction