missing translation for 'onlineSavingsMsg'
Learn More

DOG1/TMEM16A Antibody (DG1/1484), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18414593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414593 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414593 Supplier Novus Biologicals Supplier No. NBP254492AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOG1/TMEM16A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone DG1/1484
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
Host Species Mouse
Immunogen Recombinant human canine1/TMEM16A protein fragment (aa 2-101) (exact sequence is proprietary) (Uniprot: Q5XX6)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.