missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Dog (Beagle) Cryopreserved Hepatocytes, Female
Click to view available options
Quantity:
1 x 1.5mL Vial
Unit Size:
Each
Description
These cryopreserved female dog (Beagle) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.
Cells per vial: 4-8 million
Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
Phase I and II enzyme activities
Proper morphology
Viabilities generally >75%
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes
Learn about our full line of hepatocytes ,
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Cells per vial: 4-8 million
Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Learn more about our cryopreserved hepatocytes
Learn about our full line of hepatocytes ,
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Specifications
Specifications
| Content And Storage | 135°C |
| Cell Type | Hepatocytes |
| Form | Cryopreserved |
| For Use With (Application) | Cell culture, drug discovery and development, primary cell culture, hepatocyte culture, hepatocytes and hepatic models, pharma and biopharma |
| Product Type | Hepatocytes |
| Quantity | 1 x 1.5mL Vial |
| Remarks | Female Dog (Beagle) Cryopreserved Hepatocytes, 1 x 1.5mL Vial, Dog/Canine Species, Hepatocytes Cell Type, Single Donor, Liquid Nitrogen Storage, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction