missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG-1 Monoclonal Antibody (1.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512648
This item is not returnable.
View return policy
Description
MA5-12648 targets Dog1 in IHC (P, F) applications and shows reactivity with Human samples. The MA5-12648 immunogen is synthetic peptide of human Dog1.
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
Specifications
| DOG-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q5XXA6 | |
| ANO1 | |
| Synthetic peptide of human Dog1 | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 1.1 | |
| Unconjugated | |
| ANO1 | |
| ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 55107 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction