missing translation for 'onlineSavingsMsg'
Learn More

Dofetilide, Thermo Scientific Chemicals

Product Code. 16574234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16574234 1 g 1g
16584234 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16574234 Supplier Thermo Scientific Acros Supplier No. 463370010
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115256-11-6
Molecular Formula C19H27N3O5S2

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in acetone, slightly soluble in alcohol
Melting Point 149.0°C to 153.0°C
Formula Weight 441.56
Chemical Name or Material Dofetilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.