missing translation for 'onlineSavingsMsg'
Learn More

Dofetilide, MedChemExpress

Product Code. 16484307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484307 50 mg 50mg
16458428 10 mg 10mg
16450838 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484307 Supplier Medchem Express Supplier No. HYB0232
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dofetilide (UK 68789), as a class III antiarrhythmic agent, is an orally active, potent and specific IKr blocker. Dofetilide can be used for the research of cardiovascular disease.

Dofetilide (UK 68789), as a class III antiarrhythmic agent, is an orally active, potent and specific IKr blocker. Dofetilide can be used for the research of cardiovascular disease.

Specifications

Chemical Name or Material Dofetilide
CAS 115256-11-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H27N3O5S2
Quantity 50 mg
Synonym UK 68789
Solubility Information DMSO : ≥ 100 mg/mL (226.47 mM)
SMILES CS(=O)(NC1=CC=C(CCN(C)CCOC2=CC=C(NS(=O)(C)=O)C=C2)C=C1)=O
Molecular Weight (g/mol) 441.56
Formula Weight 441.56
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.