missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dofetilide
GREENER_CHOICE

Product Code. 18716688
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18716688

Brand: Tocris Bioscience™ 3757/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV11.1 (hERG) channel blocker; inhibits rapid delayed rectifier K+ current (IKr)

Dofetilide is a selective potassium channel blocker. Blocks KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Displays class III antiarrhythmic properties.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane
CAS 115256-11-6
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C19H27N3O5S2
Formula Weight Observed MW: 441.56
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.