missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodine, TRC
Specifications
Specifications
| Chemical Name or Material | Dodecylguanidinium Acetate |
| CAS | 2439-10-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 H29 N3 . C2 H4 O2 |
| Quantity | 500 mg |
| Synonym | Dodecylguanidinium Acetate, Guanidine, N-dodecyl-, acetate (1:1), Guanidine, dodecyl-, acetate (6CI,7CI), Guanidine, dodecyl-, monoacetate (8CI,9CI), AC 5223, Aadodin, American Cyanamid 5,223, Carpene, Comet, Cyprex, Cyprex 65W, Dodecylguanidine acetate, Dodecylguanidinium acetate, Dodex, Dodex (pesticide), Dodin, Dodine, Dodine 400, Dodine 544SC, Dodine 600SC, Dodine gorsac, Doguadine, Elast, Karpen, Laurylguanidine acetate, Melprex, Melprex 65, Melprex 65W, Melprex Liquid Dodine, Radspor, Syllit, Syllit 65, n-Dodecylguanidine acetate |
| InChI Formula | InChI=1S/C13H29N3.C2H4O2/c1-2-3-4-5-6-7-8-9-10-11-12-16-13(14)15;1-2(3)4/h2-12H2,1H3,(H4,14,15,16);1H3,(H,3,4) |
| SMILES | CCCCCCCCCCCCNC(=N)N.CC(=O)O |
| IUPAC Name | acetic acid;1-dodecylguanidine |
| Molecular Weight (g/mol) | 287.44 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?