missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium Chloride, TRC

Product Code. 30385359
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385359 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385359 Supplier Toronto Research Chemicals Supplier No. TRCD525500100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecyltrimethylammonium Chloride
CAS 112-00-5
Molecular Formula C15 H34 N . Cl
Quantity 100 mg
Synonym 1-Dodecanaminium, N,N,N-trimethyl-, chloride (9CI), Ammonium, dodecyltrimethyl-, chloride (8CI), Dodecyltrimethylammonium chloride (6CI), Adogen 412, Alicop, Aliquat 4, Arquad 12, Arquad 12-23, Arquad 12-25, Arquad 12-33, Arquad 12-37W, Arquad 12-50, Arquad 12D, C12TAC, CA 2150, Catinal LTC 35A, Catiogen L, Catiogen TML, Cation BB, Cation FB, Cationic 1231, Cationic Surfactant 1231, DTAC, Dehyquart LT, Emal 60W, Germicide 1231, I 0453, Laurtrimonium chloride, Lauryltrimethylammonium chloride, Lauryltrimonium chloride, Monolauryltrimethylammonium chloride, N,N,N-Trimethyl-1-dodecanaminium chloride, N-Dodecyl-N,N,N-trimethylammonium chloride, N-Lauryl-N,N,N-trimethylammonium chloride, N-Lauryltrimethylammonium chloride, Nissan Cation BB, Nissan Cation BB 300, Nissan Cation FB, Pionin B 111, Quartamin 24, Quartamin 24P, Quartamin 24W, Radiaquat 6465, Redicote E 5, Rewoquat B 18, Rhodaquat M 242C29, Swanol CA 2150, Trimethyldodecylammonium chloride, Trimethyllaurylammonium chloride, n-Dodecyltrimethylammonium chloride
InChI Formula InChI=1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)C
IUPAC Name dodecyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 263.89
Formula Weight 263.238
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.