missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium chloride, 98%

Product Code. 10586402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10586402

Brand: Thermo Scientific Acros 409320250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-00-5 | C15H34ClN | 263.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.88
MDL Number MFCD00041974
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point 237.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)11N(CH3)3Cl
Quantity 25 g
Solubility Information (10% in water) Clear
Formula Weight 263.88
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material Dodecyltrimethylammonium chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dodecyltrimethylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.