missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium Chloride, 50% Soln, MP Biomedicals

Product Code. 11411216 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411216 Supplier MP Biomedicals Supplier No. 0520448480
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cationic surfactant suitable for research and laboratory applications.

Specifications

CAS 112-00-5
Molecular Formula C15H34ClN
Quantity 100 g
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]
IUPAC Name dodecyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 263.894
PubChem CID 8152
Formula Weight 263.9
Percent Purity 50%
Physical Form Liquid
Chemical Name or Material Dodecyltrimethylammonium Chloride, 50% Soln
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.