missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium Bromide, TRC

Product Code. 30371823
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371823 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371823 Supplier Toronto Research Chemicals Supplier No. TRCD525490250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecyltrimethylammonium Bromide
CAS 1119-94-4
Purity Grade Notes HPLC
Molecular Formula C15 H34 N . Br
Quantity 250 g
Synonym N,N,N-Trimethyl-1-dodecanaminium Bromide, DDTMA, DTAB, Dctab, FSM 23, LTAB, Lauryltrimethylammonium Bromide, Morpan D, N,N,N-Trimethyl-1-dodecanaminium Bromide, N,N,N-Trimethyldodecan-1-ammonium Bromide, N-Dodecyl-N,N,N-trimethylammonium Bromide, N-Lauryl-N,N,N-trimethylammonium Bromide, Trimethyldodecylammonium Bromide, Trimethyllaurylammonium Bromide, n-Dodecyltrimethylammonium Bromide
InChI Formula InChI=1S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)C
IUPAC Name dodecyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 308.3412
Formula Weight 307.1875
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.