missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers

Product Code. 10451691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451691 500 g Glass bottle 500g
10135950 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10451691 Supplier Thermo Scientific Acros Supplier No. 325905000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85536-14-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85536-14-7
MDL Number MFCD00066677
Synonym dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid

Specifications

Melting Point -10.0°C
Color Brown
Density 1.0600g/mL
Boiling Point 315.0°C
Flash Point 210°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3(CH2)xC6H4SO3Hx=9to12
Refractive Index 1.5035 to 1.5065
Quantity 500 g
Specific Gravity 1.06
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in alkaline solutions, soluble in ethanol
Viscosity 2400 mPa.s
Percent Purity ≥96% (sulfonic acid, M.W. 322)
Physical Form Viscous Liquid
Chemical Name or Material Dodecylbenzene sulfonic acid, Mixture of C10-C13 Isomers
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.