missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene Sodium Sulfonate, 65% in water, MP Biomedicals

Product Code. 11421915 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g

Product Code. 11421915

missing translation for 'mfr': MP Biomedicals 0520331125

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 2211-98-5
Color Cream-White
Molecular Formula C18H29NaO3S
Quantity 25 g
Synonym sodium 4-dodecylbenzenesulfonate, sodium p-dodecylbenzenesulfonate, conoco c-50, unii-hb2d2zei04, hb2d2zei04, benzenesulfonic acid, 4-dodecyl-, sodium salt 1:1, dodecene-1 las, benzenesulfonic acid, 4-dodecyl-, sodium salt, sodium 4-dodecylbenzenesulphonate, 4-dodecylbenzenesulfonic acid sodium
InChI Key JHJUUEHSAZXEEO-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;4-dodecylbenzenesulfonate
Molecular Weight (g/mol) 348.477
PubChem CID 4289524
Formula Weight 348.49
Physical Form Paste
Chemical Name or Material Dodecylbenzene Sodium Sulfonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.