missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene, mixture of isomers

Product Code. p-7049928
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11357458

Brand: Thermo Scientific Alfa Aesar 031362.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-01-3 | C18H30 | 246.438 g/mol

1-Phenyldodecane acts as a pseudo-stationary phase (PSP) marker in the preparation of microemulsion, 1-Phenyldodecane was degraded to phenylacetic acid by an oil-degrading bacterium. it is used to prepare an anionic detergent dodecylbenzene sodium sulfonate. It is a petrochemical-derived substitute for the tallow and coconut oil used in making soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -3°C
Density 0.87 g/mL
Boiling Point 179°C to 180°C (13 mmHg)
Odor Specific type
Refractive Index 1.49
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 246.44
Physical Form Liquid
Chemical Name or Material Dodecylbenzene, Mixture of isomers

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.