missing translation for 'onlineSavingsMsg'
Learn More

Dodecylamine hydrochloride, 99%

Product Code. 10249890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249890 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10249890 Supplier Thermo Scientific Acros Supplier No. 409250100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 929-73-7
Melting Point 186°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C12H28ClN
Linear Formula CH3(CH2)11NH2·HCl
MDL Number MFCD00042017
Quantity 10 g
Beilstein 04, IV, 794
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform,benzene,,toluene,diethyl ether
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
SMILES [Cl-].CCCCCCCCCCCC[NH3+]
IUPAC Name dodecan-1-amine;hydrochloride
Molecular Weight (g/mol) 221.81
PubChem CID 458426
Formula Weight 221.81
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dodecylamine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dodecylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.