missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Hydrogen Maleate, TRC

Product Code. 30385903
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385903 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385903 Supplier Toronto Research Chemicals Supplier No. TRCD536823100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecyl Hydrogen Maleate
CAS 2424-61-5
Molecular Formula C16 H28 O4
Quantity 100 mg
Synonym 2-Butenedioic acid (Z)-, monododecyl ester, Maleic acid, dodecyl ester (6CI,7CI), Maleic acid, monododecyl ester (8CI), Dodecyl hydrogen maleate, Hydrogen lauryl maleate, Maleic acid monolauryl ester, Monododecyl maleate, Monolauryl maleate
InChI Formula InChI=1S/C16H28O4/c1-2-3-4-5-6-7-8-9-10-11-14-20-16(19)13-12-15(17)18/h12-13H,2-11,14H2,1H3,(H,17,18)/b13-12-
SMILES CCCCCCCCCCCCOC(=O)\C=C/C(=O)O
IUPAC Name (Z)-4-dodecoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 284.391
Formula Weight 284.199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.