missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Acrylate, TRC

Product Code. 30388058
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388058 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388058 Supplier Toronto Research Chemicals Supplier No. TRCD5253955G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylic acid-dodecyl ester
CAS 2156-97-0
Molecular Formula C15 H28 O2
Quantity 5 g
Synonym Acrylic acid, dodecyl ester (6CI,7CI,8CI), 4812/75F, Ageflex FA 12, Blemmer LA, C 12A, Dodecyl acrylate, EM 215, FA 112A, KU-LA, LA, LA 001, LAI 214, Lauryl acrylate, Light Acrylate L, Light Acrylate L-A, Miramer 120, NK Ester LA, NSC 24177, Photomer 4812, SR 335, SR 498D, Sarbio 5101, Sartomer 335, Viscoat LA, n-Dodecyl acrylate
InChI Formula InChI=1S/C15H28O2/c1-3-5-6-7-8-9-10-11-12-13-14-17-15(16)4-2/h4H,2-3,5-14H2,1H3
SMILES CCCCCCCCCCCCOC(=O)C=C
IUPAC Name dodecyl prop-2-enoate
Molecular Weight (g/mol) 240.38
Formula Weight 240.2089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.