missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoylcarnitine, MedChemExpress

Product Code. 16477487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477487 Supplier Medchem Express Supplier No. HY113166
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dodecanoylcarnitine is present in fatty acid oxidation disorders such as long-chain acyl CoA dehydrogenase deficiency, carnitine palmitoyltransferase I/II deficiency, and is also associated with celiac disease.

Dodecanoylcarnitine is present in fatty acid oxidation disorders such as long-chain acyl CoA dehydrogenase deficiency, carnitine palmitoyltransferase I/II deficiency, and is also associated with celiac disease.

Specifications

Chemical Name or Material Dodecanoylcarnitine
CAS 25518-54-1
Color White
Purity Grade Notes Research
Molecular Formula C19H37NO4
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (97.03 mM; Need ultrasonic)
SMILES CCCCCCCCCCCC(O[C@H](CC([O-])=O)C[N+](C)(C)C)=O
Molecular Weight (g/mol) 343.5
Formula Weight 343.5
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.