missing translation for 'onlineSavingsMsg'
Learn More

Dodecanophenone, 97%

Product Code. 11381477
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381477 25 g 25g
11391477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381477 Supplier Thermo Scientific Alfa Aesar Supplier No. L02014.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1674-38-0 | C18H28O | 260.421 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1674-38-0
Molecular Formula C18H28O
Molecular Weight (g/mol) 260.421
MDL Number MFCD00008967
InChI Key DJNJZIFFCJTUDS-UHFFFAOYSA-N
Synonym dodecanophenone, laurophenone, 1-dodecanone, 1-phenyl, phenyl undecyl ketone, 1-phenyl-1-dodecanone, phenyl n-undecyl ketone, n-dodecanophenone, undecyl phenyl ketone, maybridge1_003217
PubChem CID 74292
IUPAC Name 1-phenyldodecan-1-one
SMILES CCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 44°C to 46°C
Boiling Point 201°C to 202°C (9 mmHg)
Quantity 25 g
Beilstein 2050785
Formula Weight 260.42
Percent Purity 97%
Chemical Name or Material Dodecanophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.