missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoic-11,11,12,12,12-d5 Acid, CDN

Product Code. 30315114
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Product Code. Quantity unitSize
30315114 0.25 g 0.25g
30310717 0.5 g 0.5g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30315114

Brand: CDN CDND69540.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecanoic-11,11,12,12,12-D5 Acid
CAS 1219804-38-2
Molecular Formula C12 D5 H19 O2
Quantity 0.25 g
Synonym Dodecanoic-11,11,12,12,12-d5 acid (ACI), 11,11,12,12,12-Pentadeuteriododecanoic acid, Lauric-11,11,12,12,12-d5 acid, [11,11,12,12,12-2H5]Dodecanoic acid, Dodecanoic-11,11,12,12,12-D5 acid, Dodecanoic acid-11,11,12,12,12-d5, Lauric acid-11,11,12,12,12-d5
InChI Formula InChI=1 S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i1D3,2D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCC(=O)O
IUPAC Name 11,11,12,12,12-pentadeuteriododecanoic acid
Molecular Weight (g/mol) 205.3486
Formula Weight 205.209 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.