missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioyl dichloride, 98%

Product Code. 10028701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10028701 5 g Glass bottle 5g
10686671 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10028701 Supplier Thermo Scientific Acros Supplier No. 305340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4834-98-4 | C12H20Cl2O2 | 267.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4834-98-4
Molecular Formula C12H20Cl2O2
Molecular Weight (g/mol) 267.2
MDL Number MFCD00012243
InChI Key CNXXEPWXNDFGIG-UHFFFAOYSA-N
Synonym dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
PubChem CID 2733242
IUPAC Name dodecanedioyl dichloride
SMILES C(CCCCCC(=O)Cl)CCCCC(=O)Cl

Specifications

Color Yellow to Brown
Density 1.0600g/mL
Boiling Point 140°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCO(CH2)10COCl
Refractive Index 1.4670 to 1.469
Quantity 5 g
Specific Gravity 1.06
Formula Weight 267.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dodecanedioyl dichloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.