missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic acid, 99%

Product Code. 11428563
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428563 1000 g 1000g
11438563 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428563 Supplier Thermo Scientific Alfa Aesar Supplier No. A10387.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-23-2 | C12H22O4 | 230.304 g/mol

Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

Melting Point 127°C to 131°C
Density 1.15
Boiling Point 244°C to 245°C (10 mmHg)
Flash Point 227°C (440°F)
Odor Odorless
Quantity 1000 g
Beilstein 1782580
Solubility Information Soluble in water,ethanol,hot methanol,hot toluene and hot acetic acid.
Formula Weight 230.31
Percent Purity 99%
Chemical Name or Material Dodecanedioic acid
Show More Show Less
compliance-icons
Product Identifier
  • Dodecanedioic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dodecanedioic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.