missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic acid, 99%

Product Code. p-7080734
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11438563

Brand: Thermo Scientific Alfa Aesar A10387.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-23-2 | C12H22O4 | 230.304 g/mol

Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

Melting Point 127°C to 131°C
Density 1.15
Boiling Point 244°C to 245°C (10 mmHg)
Flash Point 227°C (440°F)
Odor Odorless
Quantity 250 g
Beilstein 1782580
Solubility Information Soluble in water,ethanol,hot methanol,hot toluene and hot acetic acid.
Formula Weight 230.31
Percent Purity 99%
Chemical Name or Material Dodecanedioic acid
Show More Show Less
compliance-icons
Product Identifier
  • Dodecanedioic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dodecanedioic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.