missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic Acid-13C12, TRC

Product Code. 30381769
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381769 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381769 Supplier Toronto Research Chemicals Supplier No. TRCD4945321MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecanedioic Acid-13C12
CAS 1173019-20-9
Molecular Formula 13C12H22O4
Quantity 1 mg
Synonym 1,10-Decanedicarboxylic Acid-13C12, 1,10-Dicarboxydecane-13C12, 1,12-Dodecanedioic Acid-13C12, Corfree M 2-13C12, Decamethylenedicarboxylic Acid-13C12, NSC 400242-13C12, SL-AH-13C12, n-Dodecane-alpha,ω-dioic Acid-13C12, n-Dodecanedioic Acid-13C12
InChI Formula InChI=1S/C12H22O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h1-10H2,(H,13,14)(H,15,16)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1
SMILES O[13C](=O)[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C](=O)O
IUPAC Name (1,2,3,4,5,6,7,8,9,10,11,12-^{13}C_{12})dodecanedioic acid
Molecular Weight (g/mol) 242.21
Formula Weight 242.1921
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.