missing translation for 'onlineSavingsMsg'
Learn More

Dodecanal, TRC

Product Code. 30369307
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369307 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369307 Supplier Toronto Research Chemicals Supplier No. TRCD4945452G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lauraldehyde
CAS 112-54-9
Molecular Formula C12 H24 O
Quantity 2 g
Synonym Lauraldehyde, 1-Dodecanal, Dodecanaldehyde, Dodecyl Aldehyde, Lauric Aldehyde, Laurinaldehyde, Lauryl Aldehyde, NSC 46128, NSC 52196, NSC 55212, n-Dodecanal, n-Dodecyl Aldehyde, n-Lauraldehyde
InChI Formula InChI=1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h12H,2-11H2,1H3
SMILES CCCCCCCCCCCC=O
IUPAC Name dodecanal
Molecular Weight (g/mol) 184.32
Formula Weight 184.1827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.