missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltriruthenium, 99%

Product Code. 11326008
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326008 5 g 5g
11316008 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326008 Supplier Thermo Scientific Alfa Aesar Supplier No. 010507.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carbonylation | CAS: 15243-33-1 | C12O12Ru3 | 639.33 g/mol

A H-transfer catalyst. It is used in carbonylation catalysis of the allylic amination of unactivated olefins by nitroarenes, reductive carbonylation of mononitro aromatic compounds to carbamates, and as a catalyst for cycloaddition reactions. It is employed as a precursor for the synthesis of Ru nanoparticles. Catalyst used in the intermolecular [3+2+1] cycloaddition of alpha,beta-unsaturated ketones with silylacetylenes and carbon monoxide, to produce alpha-pyrones. Catalyzes the coupling of silanes with thiols, alcohols and amines with high turnover number (TON) and high turnover frequency (TOF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15243-33-1
Molecular Formula C12O12Ru3
Molecular Weight (g/mol) 639.33
MDL Number MFCD00011209
InChI Key NQZFAUXPNWSLBI-UHFFFAOYSA-N
Synonym Ruthenium carbonyl; Triruthenium dodecacarbonyl
IUPAC Name cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1

Specifications

Melting Point ∼150°C (decomposition)
Color Orange
Linear Formula Ru3(CO)12
Quantity 5 g
UN Number UN3466
Solubility Information Sparingly soluble in hydrocarbons (e;g; hexane,cyclohexane,benzene) and acetone.
Formula Weight 639.33
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Dodecacarbonyltriruthenium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.