missing translation for 'onlineSavingsMsg'
Learn More

Docosanoic Acid Methyl Ester, TRC

Product Code. 30136833
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136833 5 g 5g
17949544 10 g 10g
17960085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136833 Supplier Toronto Research Chemicals Supplier No. TRCD6788605G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-77-1
Molecular Formula C23 H46 O2
Molecular Weight (g/mol) 354.61
Synonym Behenic Acid Methyl Ester, Kemester 9022, Kemester 9027-90, Methyl Behenate, Methyl Docosanoate, Methyl n-Docosanoate, NSC 158426
IUPAC Name methyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C23H46O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h3-22H2,1-2H3
Formula Weight 354.3498
Percent Purity >95%
Chemical Name or Material Behenic Acid Methylester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.