missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docosanedioic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Docosanedioic Acid |
| CAS | 505-56-6 |
| Molecular Formula | C22 H42 O4 |
| Quantity | 5 g |
| Synonym | 1,20-Eicosanedicarboxylic Acid, 1,22-Docosanedioic Acid, Docosan-1,22-dioic Acid, Felogenic Acid, Phellogenic Acid |
| InChI Formula | InChI=1S/C22H42O4/c23-21(24)19-17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18-20-22(25)26/h1-20H2,(H,23,24)(H,25,26) |
| SMILES | OC(=O)CCCCCCCCCCCCCCCCCCCCC(=O)O |
| IUPAC Name | docosanedioic acid |
| Molecular Weight (g/mol) | 370.567 |
| Formula Weight | 370.308 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?